Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153213.3 | NP_694945.2 | 5 | 997 | ||
XM_011540706.2 | XP_011539008.1 | 6 | 1161 | |||
GenBank mRNA | AK091281.1 | - | 5 | 1033 | ||
AK297707.1 | - | 5 | 1059 | |||
BC012982.2 | - | 4 | 573 | |||
BC040640.1 | - | 5 | 997 | 3847806 |