Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135629.2 | NP_001129101.1 | 13 | 1483 | ||
NM_001193475.1 | NP_001180404.1 | 13 | 1483 | |||
NM_152994.4 | NP_694539.1 | 13 | 1483 | |||
NR_024188.2 | 13 | 1522 | ||||
XM_017003301.1 | XP_016858790.1 | 13 | 1518 | |||
XM_017003302.1 | XP_016858791.1 | 13 | 1518 | |||
GenBank mRNA | AK122900.1 | - | 13 | 1492 | ||
AK172753.1 | - | 10 | 962 | |||
AK172762.1 | - | 13 | 1455 | |||
AK307755.1 | - | 13 | 1407 | |||
AY092062.1 | - | 13 | 1434 | |||
AY134855.1 | - | 13 | 1483 | |||
BC111059.1 | - | 13 | 1425 | 5745902 | ||
BC117279.1 | - | 13 | 1395 | 40125830 | ||
BC143463.1 | - | 13 | 1395 | 9051971 | ||
BC143466.1 | - | 13 | 1395 | 9051974 |