Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281295.1 | NP_001268224.1 | 7 | 1084 | ||
NM_001281296.1 | NP_001268225.1 | 7 | 1084 | |||
NM_001321739.1 | NP_001308668.1 | 7 | 1084 | |||
NM_138804.4 | NP_620159.2 | 7 | 1075 | |||
XM_005264152.3 | XP_005264209.1 | 3 | 244 | |||
XM_006711946.3 | XP_006712009.1 | 7 | 1420 | |||
XM_011532548.2 | XP_011530850.1 | 7 | 1421 | |||
XM_011532550.2 | XP_011530852.1 | 7 | 1031 | |||
XM_011532551.2 | XP_011530853.1 | 7 | 1030 | |||
XM_011532552.2 | XP_011530854.1 | 6 | 1247 | |||
XM_011532553.2 | XP_011530855.1 | 7 | 1422 | |||
XM_011532554.2 | XP_011530856.1 | 3 | 244 | |||
GenBank mRNA | AK126374.1 | - | 3 | 245 | ||
AK130161.1 | - | 3 | 255 | |||
AK300188.1 | - | 7 | 1076 | |||
BC014602.2 | - | 7 | 1053 | 4821635 | ||
BC025997.1 | - | 7 | 1078 | 4823281 | ||
DR158220.1 | - | 5 | 729 |