Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCIAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014446.2 | NP_001014446.1 | 2 | 253 | ||
NM_001286773.1 | NP_001273702.1 | 2 | 276 | |||
NM_001286774.1 | NP_001273703.1 | 2 | 253 | |||
NM_152398.3 | NP_689611.1 | 2 | 253 | |||
NR_104589.1 | 2 | 253 | ||||
GenBank mRNA | AK298305.1 | - | 2 | 222 | ||
AY472117.1 | - | 2 | 255 | |||
BC032808.1 | - | 2 | 224 | 4454708 | ||
BG748993.1 | - | 2 | 160 | |||
BI546814.1 | - | 2 | 258 | |||
BM756234.1 | - | 2 | 201 | |||
BM782144.1 | - | 2 | 223 | |||
BU508058.1 | - | 2 | 141 | |||
CR749310.1 | - | 2 | 198 |