Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166136.1 | NP_001159608.1 | 2 | 753 | ||
NM_147127.4 | NP_667338.3 | 2 | 315 | |||
XM_017007735.1 | XP_016863224.1 | 2 | 326 | |||
XM_017007736.1 | XP_016863225.1 | 3 | 304 | |||
XM_017007737.1 | XP_016863226.1 | 3 | 191 | |||
XM_017007738.1 | XP_016863227.1 | 2 | 326 | |||
XM_017007739.1 | XP_016863228.1 | 3 | 357 | |||
GenBank mRNA | AB083067.1 | - | 1 | 21 | ||
AY152402.1 | - | 2 | 750 | |||
AY152403.1 | - | 2 | 750 | |||
AY185210.1 | - | 2 | 315 | |||
BC143836.1 | - | 3 | 362 |