Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286668.1 | NP_001273597.1 | 14 | 2954 | ||
NM_001286669.1 | NP_001273598.1 | 14 | 3105 | |||
NM_001286670.1 | NP_001273599.1 | 12 | 2629 | |||
NM_001286671.1 | NP_001273600.1 | 12 | 2807 | |||
NM_001323890.1 | NP_001310819.1 | 16 | 3343 | |||
NM_001323892.1 | NP_001310821.1 | 14 | 3062 | |||
NM_001323893.1 | NP_001310822.1 | 13 | 2941 | |||
NM_001323899.1 | NP_001310828.1 | 14 | 3119 | |||
NM_181806.3 | NP_861522.2 | 15 | 3226 | |||
XM_017007740.1 | XP_016863229.1 | 14 | 3055 | |||
XM_017007741.1 | XP_016863230.1 | 16 | 3302 | |||
XM_017007742.1 | XP_016863231.1 | 13 | 3002 | |||
XM_017007743.1 | XP_016863232.1 | 13 | 2891 | |||
XM_017007744.1 | XP_016863233.1 | 12 | 2809 | |||
GenBank mRNA | AF516672.1 | - | 14 | 2869 | ||
AK294332.1 | - | 12 | 2591 | |||
AK303733.1 | - | 13 | 2894 | |||
AK304309.1 | - | 15 | 3217 | |||
AK316296.1 | - | 15 | 3207 | |||
AY314787.1 | - | 15 | 3189 | |||
AY422212.1 | - | 15 | 3141 | |||
BC071815.1 | - | 1 | 140 | |||
BC142709.1 | - | 12 | 2743 | 40148820 | ||
BX640635.1 | - | 10 | 4057 |