Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144645.3 | NP_653246.1 | 2 | 325 | ||
XM_011531610.2 | XP_011529912.1 | 2 | 212 | |||
XM_011531611.2 | XP_011529913.1 | 2 | 434 | |||
XM_011531612.2 | XP_011529914.1 | 2 | 144 | |||
XM_011531613.2 | XP_011529915.1 | 2 | 679 | |||
XM_011531615.2 | XP_011529917.1 | 2 | 164 | |||
XM_017007746.1 | XP_016863235.1 | 2 | 106 | |||
GenBank mRNA | BC016746.1 | - | 2 | 301 | 4827131 | |
BP289794.1 | - | 2 | 309 | |||
BX090513.1 | - | 1 | 66 |