Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 10 | 1879 | ||
NM_001289004.1 | NP_001275933.1 | 9 | 1693 | |||
NM_001308014.1 | NP_001294943.1 | 5 | 1120 | |||
NM_173488.4 | NP_775759.3 | 10 | 1879 | |||
XM_005271874.3 | XP_005271931.1 | 10 | 5076 | |||
XM_011543147.2 | XP_011541449.1 | 10 | 4971 | |||
XM_011543148.2 | XP_011541450.1 | 10 | 4839 | |||
XM_011543150.1 | XP_011541452.1 | 9 | 1089 | |||
XM_011543153.2 | XP_011541455.1 | 8 | 969 | |||
GenBank mRNA | AF505657.1 | - | 10 | 1850 | ||
AK131445.1 | - | 5 | 1017 | |||
AY273897.1 | - | 10 | 1842 | |||
BC034976.2 | - | 9 | 1695 | 4820512 |