Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130767.2 | NP_570123.1 | 15 | 1697 | ||
XM_006714531.2 | XP_006714594.1 | 14 | 1578 | |||
GenBank mRNA | AB078619.1 | - | 15 | 1665 | ||
AK122960.1 | - | 15 | 1662 | |||
BC075010.2 | - | 15 | 1663 | 30915616 | ||
BC075011.2 | - | 15 | 1658 | 30915403 |