Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301821.1 | NP_001288750.1 | 3 | 370 | ||
NM_001301822.1 | NP_001288751.1 | 3 | 370 | |||
NM_130768.2 | NP_570124.1 | 3 | 370 | |||
GenBank mRNA | AF461259.1 | - | 3 | 336 | ||
AK093445.1 | - | 3 | 323 | |||
BC028695.1 | - | 3 | 340 | |||
BC034963.1 | - | 3 | 345 | 4824818 | ||
BC126186.1 | - | 3 | 329 | 8991902 | ||
BC126188.1 | - | 3 | 329 | 8991904 | ||
BC144212.1 | - | 3 | 329 | 9052732 | ||
DB457846.1 | - | 3 | 370 |