Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145152.1 | NP_001138624.1 | 10 | 1443 | ||
NM_152415.2 | NP_689628.2 | 11 | 1518 | |||
XM_005273400.2 | XP_005273457.1 | 12 | 1528 | |||
XM_005273401.2 | XP_005273458.1 | 11 | 1474 | |||
XM_006716286.1 | XP_006716349.1 | 11 | 1465 | |||
XM_017013021.1 | XP_016868510.1 | 11 | 1483 | |||
XM_017013022.1 | XP_016868511.1 | 11 | 1483 | |||
XM_017013023.1 | XP_016868512.1 | 12 | 1510 | |||
XM_017013024.1 | XP_016868513.1 | 11 | 1456 | |||
XM_017013025.1 | XP_016868514.1 | 12 | 1510 | |||
XM_017013026.1 | XP_016868515.1 | 6 | 524 | |||
GenBank mRNA | AF547097.1 | - | 10 | 1212 | ||
AK057204.1 | - | 11 | 1439 | |||
AK124607.1 | - | 7 | 521 | |||
AY033079.2 | - | 11 | 1316 | |||
BC022363.1 | - | 7 | 674 | |||
BC067754.1 | - | 12 | 1431 | 5275060 |