Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322818.1 | NP_001309747.1 | 13 | 2144 | ||
NM_080872.3 | NP_543148.2 | 13 | 2431 | |||
XM_011544400.2 | XP_011542702.1 | 12 | 2372 | |||
XM_011544401.2 | XP_011542703.1 | 13 | 2366 | |||
XM_011544402.2 | XP_011542704.1 | 12 | 2237 | |||
XM_011544403.2 | XP_011542705.1 | 11 | 2204 | |||
XM_011544404.2 | XP_011542706.1 | 12 | 2198 | |||
XM_017013044.1 | XP_016868533.1 | 14 | 2478 | |||
XM_017013045.1 | XP_016868534.1 | 14 | 2448 | |||
XM_017013046.1 | XP_016868535.1 | 13 | 2445 | |||
XM_017013047.1 | XP_016868536.1 | 13 | 2309 | |||
XM_017013048.1 | XP_016868537.1 | 13 | 3591 | |||
XM_017013049.1 | XP_016868538.1 | 13 | 3542 | |||
XM_017013050.1 | XP_016868539.1 | 14 | 2998 | |||
GenBank mRNA | AB055056.1 | - | 13 | 2143 | ||
AK122610.1 | - | 5 | 942 | |||
AY358147.1 | - | 13 | 2431 |