Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204174.1 | NP_001191103.1 | 5 | 736 | ||
NM_001204175.1 | NP_001191104.1 | 5 | 702 | |||
NM_001204176.1 | NP_001191105.1 | 6 | 802 | |||
NM_024944.2 | NP_079220.2 | 5 | 1111 | |||
XM_011529457.2 | XP_011527759.1 | 6 | 1108 | |||
GenBank mRNA | AF257472.1 | - | 5 | 1111 | ||
AF523314.1 | - | 6 | 802 | |||
AK022689.1 | - | 6 | 984 | |||
AK296009.1 | - | 5 | 736 | |||
AK313725.1 | - | 5 | 759 | |||
AY358608.1 | - | 5 | 1097 | |||
BC009418.1 | - | 5 | 1081 | 3509743 |