Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301144.1 | NP_001288073.1 | 6 | 912 | ||
NM_001301145.1 | NP_001288074.1 | 6 | 861 | |||
NM_001301146.1 | NP_001288075.1 | 5 | 670 | |||
NM_080745.4 | NP_542783.2 | 6 | 904 | |||
NM_182985.4 | NP_892030.3 | 7 | 1381 | |||
GenBank mRNA | AF302088.1 | - | 6 | 760 | ||
AF302089.1 | - | 6 | 709 | |||
AK226115.1 | - | 5 | 5168 | |||
AK292252.1 | - | 7 | 1225 | |||
AL360161.1 | - | 6 | 747 | |||
AY305385.1 | - | 7 | 1079 | |||
BC024199.1 | - | 7 | 1199 | |||
BC033314.1 | - | 6 | 700 | 5166812 | ||
BC047945.1 | - | 7 | 1218 | 5744224 | ||
BI465168.1 | - | 5 | 500 |