Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCANP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130848.2 | NP_570900.1 | 1 | 701 | ||
GenBank mRNA | AB074498.1 | - | 1 | 701 | ||
BC069608.1 | - | 1 | 545 | 7262210 | ||
BC069610.1 | - | 1 | 545 | 7262222 | ||
BC074831.2 | - | 1 | 487 | 30915544 | ||
BC074832.2 | - | 1 | 487 | 30915326 | ||
BC117156.1 | - | 1 | 640 | |||
BC117158.1 | - | 1 | 640 |