Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290258.1 | NP_001277187.1 | 4 | 369 | ||
NM_080928.4 | NP_563616.3 | 2 | 154 | |||
XM_005250146.4 | XP_005250203.1 | 4 | 489 | |||
XM_011515816.2 | XP_011514118.1 | 2 | 150 | |||
XM_011515817.2 | XP_011514119.1 | 5 | 550 | |||
XM_011515818.2 | XP_011514120.1 | 5 | 587 | |||
XM_011515819.2 | XP_011514121.1 | 5 | 532 | |||
XM_011515820.2 | XP_011514122.1 | 3 | 364 | |||
XM_011515821.2 | XP_011514123.1 | 2 | 150 | |||
XM_017011756.1 | XP_016867245.1 | 5 | 1435 | |||
XM_017011757.1 | XP_016867246.1 | 5 | 665 | |||
GenBank mRNA | AF428257.1 | - | 2 | 154 | ||
AK125360.1 | - | 4 | 369 | |||
BC100874.2 | - | 2 | 119 | |||
BC100875.2 | - | 3 | 244 | |||
BC100876.1 | - | 3 | 244 | |||
BC100877.2 | - | 3 | 244 |