Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152433.3 | NP_689646.2 | 3 | 1762 | ||
NM_198439.2 | NP_940841.1 | 4 | 1963 | |||
XM_006718767.3 | XP_006718830.1 | 4 | 1993 | |||
XM_011542618.2 | XP_011540920.1 | 3 | 1607 | |||
XM_011542619.2 | XP_011540921.1 | 3 | 1745 | |||
GenBank mRNA | AK055247.1 | - | 1 | 379 | ||
AK092993.1 | - | 3 | 1365 | |||
AK289915.1 | - | 4 | 1573 | |||
BC117466.1 | - | 3 | 1461 | 40126017 | ||
BX640672.1 | - | 3 | 1483 | |||
BX647830.1 | - | 3 | 910 |