Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144669.2 | NP_653270.1 | 6 | 592 | ||
NR_133646.1 | 9 | 780 | ||||
XM_005253553.4 | XP_005253610.2 | 10 | 943 | |||
XM_011537956.2 | XP_011536258.2 | 11 | 991 | |||
XM_011537957.2 | XP_011536259.1 | 10 | 680 | |||
XM_017018855.1 | XP_016874344.1 | 11 | 991 | |||
XM_017018856.1 | XP_016874345.1 | 9 | 583 | |||
XM_017018857.1 | XP_016874346.1 | 8 | 556 | |||
XM_017018858.1 | XP_016874347.1 | 7 | 499 | |||
XM_017018860.1 | XP_016874349.1 | 7 | 460 | |||
XM_017018861.1 | XP_016874350.1 | 9 | 625 | |||
GenBank mRNA | AK056540.1 | - | 6 | 490 | ||
AK295333.1 | - | 11 | 708 | |||
AK295454.1 | - | 8 | 443 | |||
BC043528.1 | - | 9 | 780 | 5168267 |