Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175874.3 | NP_787070.2 | 2 | 415 | ||
XM_005253322.4 | XP_005253379.1 | 2 | 687 | |||
XM_011520568.2 | XP_011518870.1 | 2 | 1083 | |||
XM_011520569.2 | XP_011518871.1 | 2 | 259 | |||
XM_017018871.1 | XP_016874360.1 | 7 | 1126 | |||
XM_017018872.1 | XP_016874361.1 | 6 | 553 | |||
XM_017018873.1 | XP_016874362.1 | 2 | 418 | |||
XM_017018874.1 | XP_016874363.1 | 2 | 258 | |||
GenBank mRNA | AK289942.1 | - | 2 | 312 | ||
BC015546.1 | - | 2 | 287 | 3916539 | ||
BC029443.1 | - | 2 | 272 | 4670900 | ||
BC038836.1 | - | 2 | 415 | 5168757 |