Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152453.3 | NP_689666.2 | 4 | 409 | ||
XM_005254168.3 | XP_005254225.1 | 4 | 390 | |||
XM_011521262.2 | XP_011519564.1 | 4 | 390 | |||
XM_011521263.1 | XP_011519565.1 | 4 | 390 | |||
XM_011521264.1 | XP_011519566.1 | 4 | 390 | |||
GenBank mRNA | AK093126.1 | - | 4 | 390 | ||
AK310517.1 | - | 4 | 395 | |||
BC029221.2 | - | 3 | 322 | 5167874 | ||
BI826726.1 | - | 4 | 373 |