Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC47A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099646.1 | NP_001093116.1 | 16 | 1638 | ||
NM_001256663.1 | NP_001243592.1 | 17 | 1680 | |||
NM_152908.3 | NP_690872.2 | 16 | 1746 | |||
NR_135624.1 | 15 | 2133 | ||||
NR_135625.1 | 15 | 1716 | ||||
XM_017024221.1 | XP_016879710.1 | 17 | 1722 | |||
XM_017024222.1 | XP_016879711.1 | 17 | 2222 | |||
GenBank mRNA | AB250364.1 | - | 16 | 1481 | ||
AB250701.1 | - | 15 | 1559 | |||
BC035288.1 | - | 17 | 1548 | 5189385 | ||
BC050578.1 | - | 16 | 1622 | 5759988 | ||
BM930596.1 | - | 1 | 51 | |||
BX648861.1 | - | 15 | 2133 |