Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BROX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288579.1 | NP_001275508.1 | 3 | 594 | ||
NM_001288580.1 | NP_001275509.1 | 3 | 300 | |||
NM_001288581.1 | NP_001275510.1 | 3 | 300 | |||
NM_144695.3 | NP_653296.2 | 3 | 594 | |||
XM_005273065.2 | XP_005273122.2 | 3 | 230 | |||
XM_005273069.4 | XP_005273126.1 | 2 | 466 | |||
XM_006711173.3 | XP_006711236.1 | 4 | 663 | |||
XM_011509212.2 | XP_011507514.1 | 3 | 653 | |||
XM_011509213.2 | XP_011507515.1 | 2 | 538 | |||
XM_011509214.2 | XP_011507516.1 | 4 | 463 | |||
XM_017000374.1 | XP_016855863.1 | 5 | 788 | |||
XM_017000375.1 | XP_016855864.1 | 2 | 539 | |||
XM_017000376.1 | XP_016855865.1 | 3 | 591 | |||
GenBank mRNA | AB276123.1 | - | 2 | 188 | ||
AK056983.1 | - | 3 | 583 | |||
AK293974.1 | - | 3 | 226 | |||
AK304445.1 | - | 3 | 245 | |||
AK315930.1 | - | 3 | 230 | |||
BC113635.1 | - | 3 | 331 | 8322687 | ||
BC113637.1 | - | 3 | 331 | 8322689 | ||
BC143811.1 | - | 3 | 331 | 9052326 | ||
DC418391.1 | - | 3 | 300 |