Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152490.4 | NP_689703.1 | 9 | 1325 | ||
XM_006711749.3 | XP_006711812.1 | 9 | 1267 | |||
XM_017000394.1 | XP_016855883.1 | 10 | 1448 | |||
XM_017000395.1 | XP_016855884.1 | 10 | 1401 | |||
GenBank mRNA | AB209046.1 | - | 8 | 879 | ||
BC029564.1 | - | 9 | 1325 | 4826983 |