Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LCA5L SIRNA Assays ›
Gene Symbol:
LCA5L
Gene Name:
LCA5L, lebercilin like
Gene Aliases:
C21orf13
Chromosome Location:
Chr.21: 39405844 - 39445796 on Build GRCh38
UniGene ID:
Hs.517284
Species:
Human
siRNA ID
HSS135590
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LCA5L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152505.3
NP_689718.1
6
1252
XM_005260926.1
XP_005260983.1
6
1210
XM_006723967.1
XP_006724030.1
5
1255
XM_006723972.1
XP_006724035.1
6
951
XM_006723973.1
XP_006724036.1
5
791
XM_011529458.1
XP_011527760.1
8
1808
XM_011529459.1
XP_011527761.1
8
1467
XM_011529460.1
XP_011527762.1
9
1607
XM_011529461.1
XP_011527763.1
9
1714
XM_011529462.1
XP_011527764.1
7
1357
XM_011529463.2
XP_011527765.1
7
1410
XM_011529464.1
XP_011527766.1
8
1436
XM_011529465.1
XP_011527767.1
7
1355
XM_011529466.1
XP_011527768.1
7
1593
XM_011529467.1
XP_011527769.1
8
1671
XM_011529468.1
XP_011527770.1
7
1313
XM_011529469.2
XP_011527771.1
7
1700
XM_011529470.1
XP_011527772.1
6
1196
XM_017028274.1
XP_016883763.1
6
1171
XM_017028275.1
XP_016883764.1
5
924
XM_017028276.1
XP_016883765.1
6
1161
XM_017028277.1
XP_016883766.1
5
844
XM_017028278.1
XP_016883767.1
4
716
GenBank mRNA
BC031059.2
-
6
1253
5271223
BC043006.1
-
6
1469
5297703
BX648744.1
-
3
880
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0