Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK10 SIRNA Assays ›
Gene Symbol:
NEK10
Gene Name:
NIMA related kinase 10
Gene Aliases:
-
Chromosome Location:
Chr.3: 27107336 - 27369460 on Build GRCh38
UniGene ID:
Hs.506115
Species:
Human
siRNA ID
HSS135794
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031741.3
NP_001026911.1
3
317
NM_001304384.1
NP_001291313.1
3
317
NM_152534.4
NP_689747.3
25
2417
XM_006712997.3
XP_006713060.1
25
2378
XM_006712998.2
XP_006713061.1
25
2378
XM_006712999.3
XP_006713062.1
25
2378
XM_006713001.3
XP_006713064.1
25
2378
XM_011533414.2
XP_011531716.1
24
2302
XM_017005761.1
XP_016861250.1
24
2330
XM_017005762.1
XP_016861251.1
26
2494
XM_017005763.1
XP_016861252.1
25
2475
XM_017005764.1
XP_016861253.1
24
2307
XM_017005765.1
XP_016861254.1
24
2279
XM_017005766.1
XP_016861255.1
24
2330
XM_017005767.1
XP_016861256.1
23
2243
XM_017005768.1
XP_016861257.1
24
2240
XM_017005769.1
XP_016861258.1
25
2378
XM_017005770.1
XP_016861259.1
24
2330
XM_017005771.1
XP_016861260.1
24
2330
XM_017005772.1
XP_016861261.1
23
2243
XM_017005773.1
XP_016861262.1
25
2378
GenBank mRNA
AK057247.1
-
3
269
AK098832.1
-
6
500
BC045758.1
-
3
269
4821808
DB045668.1
-
3
317
DQ104438.1
-
2
40
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
microtubule-based process
chromosome segregation
positive regulation of protein autophosphorylation
positive regulation of MAP kinase activity
regulation of ERK1 and ERK2 cascade
regulation of cell cycle G2/M phase transition
protein kinase activity
protein serine/threonine kinase activity
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0