Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144719.3 | NP_653320.3 | 3 | 338 | ||
XM_005264898.3 | XP_005264955.1 | 3 | 338 | |||
XM_005264899.4 | XP_005264956.1 | 3 | 338 | |||
XM_011533418.2 | XP_011531720.1 | 3 | 338 | |||
XM_011533420.2 | XP_011531722.1 | 3 | 338 | |||
XM_011533421.2 | XP_011531723.1 | 3 | 338 | |||
GenBank mRNA | AK058196.1 | - | 3 | 338 | ||
AK302859.1 | - | 3 | 329 | |||
BC036050.1 | - | 3 | 340 | 5270468 |