Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175737.3 | NP_783864.1 | 1 | 591 | ||
GenBank mRNA | AB079373.1 | - | 1 | 591 | ||
BC104871.1 | - | 1 | 515 | 8143874 | ||
BC113653.1 | - | 1 | 515 | 8322705 | ||
BC143575.1 | - | 1 | 515 | 9052086 |