Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152542.4 | NP_689755.3 | 2 | 541 | ||
XM_005262775.3 | XP_005262832.1 | 2 | 339 | |||
XM_006714111.3 | XP_006714174.1 | 2 | 602 | |||
XM_017007803.1 | XP_016863292.1 | 2 | 189 | |||
XM_017007804.1 | XP_016863293.1 | 2 | 536 | |||
XM_017007805.1 | XP_016863294.1 | 2 | 536 | |||
GenBank mRNA | AF351614.1 | - | 2 | 272 | ||
AK054678.1 | - | 2 | 269 | |||
AK314417.1 | - | 2 | 266 | |||
AL834167.1 | - | 2 | 479 | |||
AL834271.1 | - | 2 | 260 | |||
AX766363.1 | - | 1 | 56 | |||
AY157615.1 | - | 2 | 269 | |||
AY435431.1 | - | 2 | 446 | |||
BC020850.1 | - | 2 | 271 | |||
BC037552.1 | - | 2 | 305 | 5227317 | ||
BC041350.1 | - | 1 | 1177 |