Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306149.1 | NP_001293078.1 | 2, 3 | 474 | ||
NM_001306150.1 | NP_001293079.1 | 2, 3 | 474 | |||
NM_178569.3 | NP_848664.1 | 2, 3 | 474 | |||
NR_122125.1 | 2, 3 | 416 | ||||
NR_122126.1 | 2, 3 | 416 | ||||
GenBank mRNA | AY249325.1 | - | 2, 3 | 457 | ||
BC101608.1 | - | 2, 3 | 377 | 8069114 | ||
BC101634.1 | - | 2, 3 | 377 | 8069140 | ||
BC143638.1 | - | 2, 3 | 319 | |||
BC143639.1 | - | 2, 3 | 377 | 9052153 | ||
BC143640.1 | - | 2, 3 | 319 | |||
CX754076.1 | - | 2, 3 | 359 |