Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134405.1 | NP_001127877.1 | 3 | 661 | ||
NM_001134406.1 | NP_001127878.1 | 3 | 661 | |||
NM_138290.2 | NP_612147.1 | 4 | 712 | |||
XM_005250156.2 | XP_005250213.1 | 3 | 661 | |||
XM_011515826.1 | XP_011514128.1 | 4 | 712 | |||
XM_011515827.1 | XP_011514129.1 | 4 | 712 | |||
XM_011515828.2 | XP_011514130.1 | 4 | 195 | |||
XM_011515829.2 | XP_011514131.1 | 3 | 329 | |||
XM_017011761.1 | XP_016867250.1 | 4 | 399 | |||
XM_017011762.1 | XP_016867251.1 | 5 | 324 | |||
XM_017011763.1 | XP_016867252.1 | 2 | 138 | |||
XM_017011765.1 | XP_016867254.1 | 4 | 311 | |||
XM_017011766.1 | XP_016867255.1 | 4 | 590 | |||
GenBank mRNA | AK055233.1 | - | 4 | 710 | ||
AK293607.1 | - | 3 | 477 | |||
AK294039.1 | - | 3 | 459 | |||
BC022520.1 | - | 3 | 612 | 4793693 | ||
DC304423.1 | - | 3 | 477 |
Gene Symbol: ADAM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DR000609.1 | - | 589 |