Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201286.3 | NP_958443.1 | 2 | 1644 | ||
XM_017029299.1 | XP_016884788.1 | 3 | 1911 | |||
XM_017029300.1 | XP_016884789.1 | 3 | 1794 | |||
XM_017029301.1 | XP_016884790.1 | 2 | 1714 | |||
GenBank mRNA | AJ583823.2 | - | 1 | 1565 |