Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178499.4 | NP_848594.2 | 6 | 1039 | ||
XM_017018911.1 | XP_016874400.1 | 5 | 714 | |||
XM_017018912.1 | XP_016874401.1 | 6 | 822 | |||
XM_017018913.1 | XP_016874402.1 | 6 | 822 | |||
XM_017018914.1 | XP_016874403.1 | 2 | 105 | |||
GenBank mRNA | BC040553.1 | - | 6 | 1039 | 5301593 | |
BM724243.1 | - | 4 | 354 |