Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LGI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139284.2 | NP_644813.1 | 9 | 2175 | ||
XM_011526595.1 | XP_011524897.1 | 10 | 1656 | |||
XM_017026428.1 | XP_016881917.1 | 9 | 1504 | |||
XM_017026429.1 | XP_016881918.1 | 10 | 1576 | |||
XM_017026430.1 | XP_016881919.1 | 8 | 1358 | |||
GenBank mRNA | AF467954.1 | - | 9 | 1867 | ||
AJ487519.1 | - | 9 | 2100 | |||
AJ487959.1 | - | 9 | 1608 | |||
AL832968.1 | - | 6 | 2076 | |||
BC029943.1 | - | 10 | 1640 | |||
BC136694.1 | - | 9 | 1650 | 9020684 | ||
BC136697.1 | - | 9 | 1650 | 9020687 |