Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198621.2 | NP_001185550.1 | 3 | 4881 | ||
NM_194293.2 | NP_919269.2 | 2 | 5298 | |||
XM_005264909.3 | XP_005264966.1 | 2 | 5257 | |||
GenBank mRNA | AJ271461.1 | - | 1 | 426 | ||
AJ626899.1 | - | 3 | 4879 | |||
AJ626900.1 | - | 2 | 5306 | |||
AK095941.1 | - | 1 | 921 | |||
AK096421.1 | - | 2 | 1443 | |||
AK122723.1 | - | 2 | 2278 | |||
AK126299.1 | - | 1 | 3748 | |||
AL713648.1 | - | 1 | 3386 | |||
AL831956.1 | - | 2 | 2666 | |||
AL832020.1 | - | 3 | 2917 | |||
AL832459.1 | - | 2 | 2959 | |||
AY375160.1 | - | 2 | 5298 | |||
BC127119.1 | - | 2 | 1429 | |||
BC139782.1 | - | 2 | 1429 | |||
CR749430.1 | - | 3 | 4881 |