Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153361.3 | NP_699192.1 | 4 | 1776 | ||
NR_072980.1 | 4 | 2237 | ||||
XM_006714450.1 | XP_006714513.1 | 4 | 1563 | |||
XM_006714451.1 | XP_006714514.1 | 3 | 804 | |||
XM_017009164.1 | XP_016864653.1 | 4 | 1205 | |||
XM_017009165.1 | XP_016864654.1 | 4 | 978 | |||
GenBank mRNA | AK122985.1 | - | 4 | 2237 | ||
BC036422.2 | - | 4 | 1619 | 4797792 |