Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122769.2 | NP_001116241.1 | 8 | 2336 | ||
NM_181714.3 | NP_859065.2 | 9 | 2491 | |||
XM_005248665.4 | XP_005248722.1 | 8 | 2350 | |||
XM_011535504.1 | XP_011533806.1 | 9 | 2403 | |||
GenBank mRNA | AL832214.1 | - | 2 | 795 | ||
BC050327.1 | - | 9 | 2491 | 4823983 | ||
BX648161.1 | - | 8 | 2317 |