Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139056.3 | NP_620687.2 | 5 | 956 | ||
NR_136935.1 | 5 | 956 | ||||
GenBank mRNA | AB095949.1 | - | 2 | 211 | ||
AJ315734.1 | - | 5 | 818 | |||
AK122980.1 | - | 3 | 581 | |||
AK127746.1 | - | 5 | 956 | |||
DQ266047.1 | - | 5 | 956 |