Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128933.2 | NP_001122405.1 | 4 | 3374 | ||
NM_001128934.2 | NP_001122406.1 | 4 | 3374 | |||
NM_001286754.1 | NP_001273683.1 | 4 | 3326 | |||
NM_133477.2 | NP_597734.2 | 4 | 3374 | |||
GenBank mRNA | AF177291.1 | - | 1 | 299 | ||
AJ010482.1 | - | 2 | 2394 | |||
AK296962.1 | - | 2 | 2140 | |||
AL832031.1 | - | 4 | 3357 | |||
AL832363.1 | - | 4 | 3268 | |||
AL833294.1 | - | 4 | 3360 | |||
AL833547.1 | - | 4 | 3374 | |||
BC136256.1 | - | 4 | 3249 | 9020243 | ||
BC150629.1 | - | 4 | 3373 | 9056999 | ||
BX647849.1 | - | 2 | 2883 | |||
CR936673.1 | - | 4 | 3366 | |||
EU481975.1 | - | 4 | 3178 | |||
FN422000.1 | - | 2 | 2429 | |||
KF147165.1 | - | 4 | 3326 |