Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 6 | 689 | ||
XM_006719272.3 | XP_006719335.1 | 6 | 843 | |||
GenBank mRNA | AL832200.1 | - | 2 | 242 | ||
AY727870.2 | - | 6 | 689 | |||
CN419675.1 | - | 2 | 153 | |||
CR749833.1 | - | 5 | 608 | |||
DQ096628.1 | - | 6 | 689 |