Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152647.2 | NP_689860.2 | 15 | 1696 | ||
XM_005254213.3 | XP_005254270.1 | 16 | 1848 | |||
XM_005254214.3 | XP_005254271.1 | 16 | 1781 | |||
XM_005254215.3 | XP_005254272.1 | 15 | 1812 | |||
XM_005254216.3 | XP_005254273.1 | 15 | 1746 | |||
XM_005254219.3 | XP_005254276.1 | 13 | 1534 | |||
XM_006720423.2 | XP_006720486.1 | 17 | 1813 | |||
XM_006720424.2 | XP_006720487.1 | 15 | 1773 | |||
XM_006720426.2 | XP_006720489.1 | 14 | 1707 | |||
XM_006720427.3 | XP_006720490.1 | 17 | 1819 | |||
XM_011521319.2 | XP_011519621.1 | 16 | 3731 | |||
XM_011521320.1 | XP_011519622.1 | 14 | 1223 | |||
XM_011521322.1 | XP_011519624.1 | 13 | 1187 | |||
XM_017021990.1 | XP_016877479.1 | 13 | 1605 | |||
XM_017021991.1 | XP_016877480.1 | 17 | 1888 | |||
XM_017021992.1 | XP_016877481.1 | 14 | 1536 | |||
XM_017021993.1 | XP_016877482.1 | 12 | 1530 | |||
XM_017021994.1 | XP_016877483.1 | 12 | 1506 | |||
XM_017021995.1 | XP_016877484.1 | 14 | 1589 | |||
XM_017021996.1 | XP_016877485.1 | 15 | 1296 | |||
GenBank mRNA | AK023699.1 | - | 2 | 137 | ||
AK057362.1 | - | 15 | 1694 | |||
BG182490.1 | - | 2 | 310 |
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720461.3 | XP_006720524.1 | 11 | 4572 | ||
XM_017022066.1 | XP_016877555.1 | 12 | 4849 |