Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001214906.1 | NP_001201835.1 | 3 | 1790 | ||
NM_001214907.1 | NP_001201836.1 | 2 | 1694 | |||
NM_001214909.1 | NP_001201838.1 | 3 | 2158 | |||
NM_001324494.1 | NP_001311423.1 | 2 | 2064 | |||
NM_152652.2 | NP_689865.2 | 2 | 2052 | |||
GenBank mRNA | AK056313.1 | - | 2 | 1735 | ||
AK122598.1 | - | 1 | 819 | |||
AL832433.1 | - | 1 | 735 | |||
BC007393.2 | - | 1 | 321 |