Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105513.2 | NP_001098983.2 | 10 | 1403 | ||
NM_145755.2 | NP_665698.2 | 11 | 1550 | |||
XM_005264921.4 | XP_005264978.1 | 11 | 1476 | |||
XM_005264922.4 | XP_005264979.1 | 11 | 1476 | |||
XM_005264923.3 | XP_005264980.1 | 11 | 1476 | |||
XM_005264924.4 | XP_005264981.1 | 11 | 1476 | |||
XM_005264925.4 | XP_005264982.1 | 10 | 1353 | |||
XM_005264926.4 | XP_005264983.1 | 11 | 1476 | |||
XM_005264927.4 | XP_005264984.1 | 7 | 930 | |||
XM_006713011.3 | XP_006713074.1 | 11 | 1476 | |||
XM_011533447.2 | XP_011531749.1 | 10 | 1377 | |||
XM_011533449.1 | XP_011531751.1 | 11 | 1476 | |||
XM_017005843.1 | XP_016861332.1 | 11 | 1476 | |||
GenBank mRNA | AJ487015.1 | - | 11 | 1512 | ||
AK093313.1 | - | 1 | 87 | |||
AK302408.1 | - | 10 | 1386 | |||
AK302465.1 | - | 10 | 1257 | |||
BC129948.1 | - | 11 | 1458 | 40126472 |