Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168364.1 | NP_001161836.1 | 4, 5 | 524 | ||
NM_001321325.1 | NP_001308254.1 | 4, 5 | 524 | |||
NM_173853.3 | NP_776252.2 | 4, 5 | 524 | |||
GenBank mRNA | AY157576.1 | - | 4, 5 | 494 | ||
AY358993.1 | - | 3, 4 | 582 | |||
BC101687.1 | - | 4, 5 | 523 | 8069193 | ||
BC101689.1 | - | 4, 5 | 524 | 8069195 | ||
BC143574.1 | - | 4, 5 | 523 | 9052085 | ||
CB160871.1 | - | 3, 4 | 455 |