Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145702.2 | NP_663748.1 | 1 | 1045 | ||
GenBank mRNA | AK056329.1 | - | 1 | 971 | ||
BC035143.1 | - | 1 | 1050 | 5264460 | ||
BC063500.1 | - | 1 | 1005 | 5248661 |