Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198463.2 | NP_940865.1 | 15 | 1959 | ||
XM_005264929.2 | XP_005264986.1 | 16 | 2678 | |||
XM_006713013.3 | XP_006713076.1 | 16 | 2717 | |||
XM_011533451.2 | XP_011531753.1 | 15 | 2552 | |||
XM_011533453.2 | XP_011531755.1 | 15 | 2655 | |||
XM_011533456.1 | XP_011531758.1 | 16 | 2426 | |||
XM_011533457.1 | XP_011531759.1 | 14 | 2206 | |||
XM_011533458.2 | XP_011531760.1 | 13 | 2089 | |||
XM_011533459.2 | XP_011531761.1 | 15 | 2339 | |||
XM_011533460.2 | XP_011531762.1 | 11 | 1856 | |||
XM_011533461.1 | XP_011531763.1 | 13 | 2049 | |||
XM_011533463.2 | XP_011531765.1 | 10 | 1680 | |||
XM_017005848.1 | XP_016861337.1 | 12 | 1962 | |||
XM_017005849.1 | XP_016861338.1 | 15 | 2560 | |||
XM_017005850.1 | XP_016861339.1 | 14 | 1860 | |||
GenBank mRNA | AK124111.1 | - | 15 | 1961 | ||
AK124920.1 | - | 8 | 1429 | |||
AK128200.1 | - | 4 | 527 | |||
BC050317.1 | - | 15 | 2662 | |||
BC132815.1 | - | 13 | 1773 | 40146837 | ||
BC151142.1 | - | 13 | 1773 | 9020498 |