Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282506.1 | NP_001269435.1 | 2 | 1893 | ||
NM_001291833.1 | NP_001278762.1 | 3 | 1368 | |||
NM_152673.3 | NP_689886.3 | 3 | 1380 | |||
GenBank mRNA | AB037780.1 | - | 2 | 3189 | ||
AB098731.1 | - | 2 | 1631 | |||
AK027314.1 | - | 4 | 1131 | |||
AK075138.1 | - | 3 | 1089 | |||
AK311504.1 | - | 3 | 1378 | |||
AY358449.1 | - | 3 | 1313 | |||
BC044243.1 | - | 3 | 1267 | 5754328 | ||
BF883124.1 | - | 1 | 200 | |||
BM697386.1 | - | 1 | 480 |