Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282290.1 | NP_001269219.1 | 10 | 1824 | ||
NM_199282.2 | NP_954976.1 | 10 | 1232 | |||
XM_005257128.3 | XP_005257185.1 | 9 | 1281 | |||
XM_006721745.2 | XP_006721808.1 | 13 | 2206 | |||
XM_006721747.2 | XP_006721810.1 | 12 | 2140 | |||
XM_006721750.3 | XP_006721813.1 | 13 | 2179 | |||
XM_011524468.1 | XP_011522770.1 | 13 | 2206 | |||
XM_011524469.1 | XP_011522771.1 | 12 | 2140 | |||
XM_011524470.2 | XP_011522772.1 | 13 | 2179 | |||
XM_011524471.1 | XP_011522773.1 | 13 | 2035 | |||
XM_011524472.2 | XP_011522774.1 | 10 | 1347 | |||
XM_011524474.1 | XP_011522776.1 | 10 | 1217 | |||
XM_017024311.1 | XP_016879800.1 | 11 | 2140 | |||
XM_017024312.1 | XP_016879801.1 | 10 | 1345 | |||
XM_017024313.1 | XP_016879802.1 | 13 | 2179 | |||
GenBank mRNA | AK125535.1 | - | 10 | 1217 | ||
AK300880.1 | - | 9 | 1238 | |||
BC101388.1 | - | 9 | 1000 | 40026573 | ||
BC101389.1 | - | 9 | 1000 | 40026575 | ||
BC101390.1 | - | 9 | 1000 | 40026576 | ||
BC101391.2 | - | 8 | 919 | 40026577 |