Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282290.1 | NP_001269219.1 | 14 | 2243 | ||
NM_199282.2 | NP_954976.1 | 14 | 1651 | |||
XM_005257128.3 | XP_005257185.1 | 13 | 1700 | |||
XM_006721745.2 | XP_006721808.1 | 17 | 2625 | |||
XM_006721747.2 | XP_006721810.1 | 16 | 2559 | |||
XM_011524468.1 | XP_011522770.1 | 18 | 2724 | |||
XM_011524469.1 | XP_011522771.1 | 17 | 2658 | |||
XM_011524470.2 | XP_011522772.1 | 18 | 2697 | |||
XM_011524471.1 | XP_011522773.1 | 18 | 2553 | |||
XM_011524472.2 | XP_011522774.1 | 15 | 1865 | |||
XM_011524474.1 | XP_011522776.1 | 15 | 1735 | |||
XM_017024311.1 | XP_016879800.1 | 16 | 2658 | |||
XM_017024312.1 | XP_016879801.1 | 14 | 1764 | |||
GenBank mRNA | AK125535.1 | - | 14 | 1636 | ||
BC101388.1 | - | 13 | 1419 | 40026573 | ||
BC101389.1 | - | 13 | 1419 | 40026575 | ||
BC101390.1 | - | 13 | 1419 | 40026576 | ||
BC101391.2 | - | 12 | 1338 | 40026577 |