Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174922.4 | NP_777582.4 | 14 | 1690 | ||
XM_006716527.2 | XP_006716590.1 | 14 | 1680 | |||
XM_011516907.2 | XP_011515209.1 | 14 | 2403 | |||
XM_011516909.1 | XP_011515211.1 | 14 | 1690 | |||
XM_011516910.1 | XP_011515212.1 | 15 | 1713 | |||
XM_017013174.1 | XP_016868663.1 | 14 | 1728 | |||
GenBank mRNA | AK092773.1 | - | 14 | 1653 | ||
BC031570.1 | - | 3 | 423 | |||
BC064345.1 | - | 5 | 641 | |||
BC085013.1 | - | 9 | 1000 | |||
BC101659.1 | - | 14 | 1630 | 8069165 | ||
BC143326.1 | - | 13 | 1457 | |||
BI825038.1 | - | 3 | 453 |