Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153045.3 | NP_694590.2 | 9 | 1163 | ||
XM_006716997.1 | XP_006717060.1 | 9 | 1166 | |||
XM_011518350.1 | XP_011516652.1 | 10 | 1334 | |||
XM_011518351.2 | XP_011516653.1 | 10 | 1462 | |||
XM_011518352.1 | XP_011516654.1 | 10 | 1331 | |||
XM_011518353.1 | XP_011516655.1 | 9 | 1223 | |||
XM_011518354.1 | XP_011516656.1 | 9 | 1369 | |||
XM_011518355.1 | XP_011516657.1 | 9 | 1366 | |||
XM_017014429.1 | XP_016869918.1 | 10 | 1490 | |||
XM_017014430.1 | XP_016869919.1 | 8 | 1255 | |||
GenBank mRNA | AK095364.1 | - | 4 | 487 | ||
AK289383.1 | - | 9 | 1132 | |||
AL834412.1 | - | 7 | 775 | |||
BC019913.1 | - | 3 | 336 | |||
BC062550.1 | - | 5 | 505 | |||
BC107133.1 | - | 9 | 1101 | 40025594 | ||
BC107134.1 | - | 9 | 1314 | 40025600 | ||
BX649023.1 | - | 9 | 1049 |